Ethyl-2,4,6-trimethylphenylacetate 97%
Supplier: Thermo Scientific Chemicals
L08275-04
L08275-09
CAAAAL08275-04EA
93.06
CAD
CAAAAL08275-04
CAAAAL08275-09
Ethyl-2,4,6-trimethylphenylacetate 97%
Ethyl-2,4,6-trimethylphenylacetate
Purity: 97%
CAS No.: 5460-08-2
Molecular Formula: C13H18O2
Formula Weight: 206.29
Refractive Index: 1.5050
Boiling Point: 135-140°/9mm
Beilstein Reference: 2105791
MDL No.: MFCD00051916
Formula:
C₁₃H₁₈O₂ Boiling Pt: 135…140 °C/9 mm Density: 1.034 Storage Temperature: Ambient |
MDL Number:
MFCD00051916 CAS Number: 5460-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...