Lithium aluminium di-n-butylamide solution_SC 0.16 M 1,2-Dimethoxyethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithium tetrakis(N-butyl-1-butanaminato)aluminate(1-), Aluminate(1-), tetrakis(N-butyl-1-butanaminato)-, lithium (1:1)
L08269-AE
L08269-AP
CAAAAL08269-AEEA
278.08
CAD
CAAAAL08269-AE
CAAAAL08269-AP
Lithium aluminium di-n-butylamide solution_SC 0.16 M 1,2-Dimethoxyethane
Lithium aluminium di-n-butylamide
CAS No.: 15405-86-4
Molecular Formula: C32H72AlLiN4
Formula Weight: 546.87
Storage Temperature: 2°C to 8°C
Melting Point: -54°
UN No.: 1993
MDL No.: MFCD00051918
Formula:
C₃₂H₇₂AlLiN₄ MW: 546.87 g/mol Melting Pt: –54 °C Density: 0.860 Storage Temperature: Ambient |
MDL Number:
MFCD00051918 CAS Number: 15405-86-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...