4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) predominantly trans ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(tert-butyl)cyclohexanecarboxylic acid
L08248-06
L08248-14
CAAAAL08248-06EA
94.33
CAD
CAAAAL08248-06
CAAAAL08248-14
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) predominantly trans ≥98%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Purity: 98+%
CAS No.: 5451-55-8
Molecular Formula: C11H20O2
Formula Weight: 184.28
Beilstein Reference: 2043187
MDL No.: MFCD00042622
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00042622 CAS Number: 5451-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...