tert-Butyl 2-bromoisobutyrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl α-bromoisobutyrate, 2-Bromoisobutyric acid tert-butyl ester, tert-Butyl 2-methyl-2-propionate, tert-Butyl 2-bromo-2-methylpropanoate
L08216-09
L08216-18
L08216-30
CAAAAL08216-09EA
47.56
CAD
CAAAAL08216-09
CAAAAL08216-18
CAAAAL08216-30
tert-Butyl 2-bromoisobutyrate 97%
tert-Butyl 2-bromoisobutyrate
Purity: 97%
CAS No.: 23877-12-5
Molecular Formula: C8H15BrO2
Formula Weight: 223.12
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4360
Boiling Point: 35-37°/1mm
Beilstein Reference: 1905654
MDL No.: MFCD00051908
Formula:
C₆H₁₁BrO₂ MW: 223.12 g/mol Boiling Pt: 35…37 °C/1 mm Density: 1.180 Storage Temperature: Refrigerator |
MDL Number:
MFCD00051908 CAS Number: 23877-12-5 EINECS: 245-924-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...