5-Methoxypiperonal 97%

Supplier: Thermo Scientific Chemicals

L08213-03 L08213-06 L08213-14
CAAAAL08213-03EA 82.63 CAD
CAAAAL08213-03 CAAAAL08213-06 CAAAAL08213-14
5-Methoxypiperonal 97%
5-Methoxypiperonal

Purity: 97% CAS No.: 5780-07-4 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 126-132° Beilstein Reference: 384077

Formula: C₉H₈O₄
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00016900
CAS Number: 5780-07-4
EINECS: 227-304-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR