2-(1-Pyrrolidinyl)phenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08178-03
L08178-06
CAAAAL08178-03EA
106.81
CAD
CAAAAL08178-03
CAAAAL08178-06
2-(1-Pyrrolidinyl)phenol 98%
2-(1-Pyrrolidinyl)phenol
Purity: 98%
CAS No.: 4787-77-3
Molecular Formula: C10H13NO
Formula Weight: 163.22
Melting Point: 109-112°
Beilstein Reference: 135437
Formula:
C₁₀H₁₃NO MW: 163.22 g/mol Melting Pt: 109…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051659 CAS Number: 4787-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...