Tetra-n-dodecylammonium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

L08153-04 L08153-09
CAAAAL08153-04EA 83.42 CAD
CAAAAL08153-04 CAAAAL08153-09
Tetra-n-dodecylammonium iodide 98%
Tetra-n-dodecylammonium iodide

Purity: 98% CAS No.: 40797-39-5 Molecular Formula: C48H100IN Formula Weight: 818.23 Melting Point: 114-118° Beilstein Reference: 5844188 MDL No.: MFCD00051856

Formula: C₄₈H₁₀₀IN
MW: 818.23 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00051856
CAS Number: 40797-39-5
EINECS: 255-086-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR