Methyl-2,5-dichlorothiophene-3-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08118-03
L08118-06
CAAAAL08118-03EA
108.39
CAD
CAAAAL08118-03
CAAAAL08118-06
Methyl-2,5-dichlorothiophene-3-carboxylate 98%
Methyl-2,5-dichlorothiophene-3-carboxylate
Purity: 98%
CAS No.: 145129-54-0
Molecular Formula: C6H4Cl2O2S
Formula Weight: 211.07
Refractive Index: 1.5630
Boiling Point: 74-75°/0.6mm
MDL No.: MFCD00051763
Formula:
C₆H₄Cl₂O₂S MW: 211.07 g/mol Boiling Pt: 74…75 °C/0.6 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051763 CAS Number: 145129-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...