Methyl methanesulfonylacetate ≥98%
Supplier: Thermo Scientific Chemicals
L08039-06
L08039-14
CAAAAL08039-06EA
116.13
CAD
CAAAAL08039-06
CAAAAL08039-14
Methyl methanesulfonylacetate ≥98%
Methyl methanesulfonylacetate
Purity: 98+%
CAS No.: 62020-09-1
Molecular Formula: C4H8O4S
Formula Weight: 152.17
Melting Point: 59-63°
Beilstein Reference: 2412782
MDL No.: MFCD00051842
Formula:
C₄H₈O₄S Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051842 CAS Number: 62020-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...