Ethyl-3-(4-methoxybenzoyl)propionate 98%
Supplier: Thermo Scientific Chemicals
L07989-06
L07989-14
CAAAAL07989-06EA
111.07
CAD
CAAAAL07989-06
CAAAAL07989-14
Ethyl-3-(4-methoxybenzoyl)propionate 98%
Ethyl-3-(4-methoxybenzoyl)propionate
Purity: 98%
CAS No.: 15118-67-9
Molecular Formula: C13H16O4
Formula Weight: 236.27
Melting Point: 49-53°
Beilstein Reference: 2120509
MDL No.: MFCD00051784
Formula:
C₁₃H₁₆O₄ Melting Pt: 49…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051784 CAS Number: 15118-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...