4-Dimethylamino-1-naphthoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07944-03
L07944-06
CAAAAL07944-03EA
199.08
CAD
CAAAAL07944-03
CAAAAL07944-06
CAAAAL07944-MD
4-Dimethylamino-1-naphthoic acid 98%
4-Dimethylamino-1-naphthoic acid
Purity: 98%
CAS No.: 78062-03-0
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Melting Point: ca 160° dec.
Beilstein Reference: 2723206
MDL No.: MFCD00051797
Formula:
C₁₃H₁₃NO₂ Melting Pt: ca 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051797 CAS Number: 78062-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...