3-(4-Hydroxyphenyl)propiononitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4-Hydroxyphenyl)propionitrile
L07934-03
L07934-06
CAAAAL07934-03EA
64.31
CAD
CAAAAL07934-03
CAAAAL07934-06
3-(4-Hydroxyphenyl)propiononitrile 98%
3-(4-Hydroxyphenyl)propiononitrile
Purity: 98%
CAS No.: 17362-17-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 54-58°
Boiling Point: 166°/8mm
UN No.: 3439
MDL No.: MFCD00016492
Formula:
C₉H₉NO MW: 147.18 g/mol Boiling Pt: 166 °C/8 mm Melting Pt: 54…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016492 CAS Number: 17362-17-3 EINECS: 241-393-0 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...