Ethyl-1-naphthoate 97%
Supplier: Thermo Scientific Chemicals
L07760-09
L07760-18
CAAAAL07760-09EA
76.31
CAD
CAAAAL07760-09
CAAAAL07760-18
Ethyl-1-naphthoate 97%
Ethyl 1-naphthoate
Purity: 97%
CAS No.: 3007-97-4
Molecular Formula: C13H12O2
Formula Weight: 200.24
Refractive Index: 1.5930
Boiling Point: 308-310°
MDL No.: MFCD00016805
Formula:
C₁₃H₁₂O₂ Boiling Pt: 308…310 °C Density: 1,110 Storage Temperature: Ambient |
MDL Number:
MFCD00016805 CAS Number: 3007-97-4 EINECS: 221-120-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...