3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,3',5,5'-Tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione, 3,3',5,5'-Tetra-tert-butyldiphenoquinone
L07743-04
L07743-09
CAAAAL07743-04EA
184.86
CAD
CAAAAL07743-04
CAAAAL07743-09
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone
Purity: 98+%
CAS No.: 2455-14-3
Molecular Formula: C28H40O2
Formula Weight: 408.63
Melting Point: 242-246°
Beilstein Reference: 1916759
MDL No.: MFCD00051798
Formula:
C₂₈H₄₀O₂ Melting Pt: 242…246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051798 CAS Number: 2455-14-3 EINECS: 219-527-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...