1-Hexen-3-one ≥90% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 0.5%
L07660-06
L07660-14
CAAAAL07660-06EA
200.66
CAD
CAAAAL07660-06
CAAAAL07660-14
1-Hexen-3-one ≥90% stabilized
1-Hexen-3-one
Stab. with 0.5% 4-methoxyphenol
Purity: 90+%
CAS No.: 1629-60-3
Molecular Formula: C6H10O
Formula Weight: 98.15
Storage Temperature: 2°C to 8°C
Flash Point: 22°(71°F)
Refractive Index: 1.4275
Odor: Pungent
Boiling Point: 127-129°
Beilstein Reference: 1740796
UN No.: 1224
MDL No.: MFCD00051563
Formula:
C₆H₁₀O Boiling Pt: 127…129 °C Density: 0.840 Flash Pt: 22 °C(71 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051563 CAS Number: 1629-60-3 EINECS: 216-625-9 UN: 1224 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...