4-Fluoro-DL-phenylalanine 98+%
Supplier: Thermo Scientific Chemicals
L07585-03
L07585-06
CAAAAL07585-03EA
72.68
CAD
CAAAAL07585-03
CAAAAL07585-06
4-Fluoro-DL-phenylalanine 98+%
4-Fluoro-DL-phenylalanine
Purity: 98+%
CAS No.: 51-65-0
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Melting Point: ca 254° dec.
Beilstein Reference: 29338793
Formula:
C₉H₁₀FNO₂ Melting Pt: ca 254 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002600 CAS Number: 51-65-0 EINECS: 200-113-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...