Quinoxaline-2,3-dione 98%

Supplier: Thermo Scientific Chemicals

L07581-14 L07581-22
CAAAAL07581-14EA 102.23 CAD
CAAAAL07581-14 CAAAAL07581-22
Quinoxaline-2,3-dione 98%
Quinoxaline-2,3-dione

Purity: 98% CAS No.: 15804-19-0 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 300° Beilstein Reference: 136884

Formula: C₈H₆N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006723
CAS Number: 15804-19-0
EINECS: 239-901-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR