Ethyl-4-nitrophenylglyoxylate 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-(4-nitrophenyl)-2-oxoacetate
L07579-03
L07579-06
CAAAAL07579-03EA
120.08
CAD
CAAAAL07579-03
CAAAAL07579-06
Ethyl-4-nitrophenylglyoxylate 98+%
Ethyl 4-nitrophenylglyoxylate
Purity: 98+%
CAS No.: 70091-75-7
Molecular Formula: C10H9NO5
Formula Weight: 223.19
Melting Point: 39-43°
Boiling Point: 152-157°/0.5mm
MDL No.: MFCD00051841
Formula:
C₁₀H₉NO₅ MW: 223.19 g/mol Boiling Pt: 152…157 °C/0.5 mm Melting Pt: 39…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051841 CAS Number: 70091-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...