Acetazolamide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Acetamido-5-sulphonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulphonamide, 2-Acetamido-5-sulfonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide
L07562-06
L07562-14
CAAAAL07562-06EA
60.04
CAD
CAAAAL07562-06
CAAAAL07562-14
Acetazolamide 99%
Acetazolamide
Carbonic anhydrase inhibitor; increases cerebral blood flow
Purity: 99%
CAS No.: 59-66-5
Molecular Formula: C4H6N4O3S2
Formula Weight: 222.25
Melting Point: ca 255° dec.
Beilstein Reference: 212994
MDL No.: MFCD00003105
Formula:
C₄H₆N₄O₃S₂ MW: 222.24 g/mol Melting Pt: ca 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003105 CAS Number: 59-66-5 EINECS: 200-440-5 Merck Index: 14,53 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...