2-(Trimethylacetyl)thiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Dimethyl-1-(thiophen-2-yl)propan-1-one
L07560-06
L07560-14
CAAAAL07560-06EA
125.93
CAD
CAAAAL07560-06
CAAAAL07560-14
2-(Trimethylacetyl)thiophene 98%
2-(Trimethylacetyl)thiophene
Purity: 98%
CAS No.: 20409-48-7
Molecular Formula: C9H12OS
Formula Weight: 168.26
Flash Point: 99°(210°F)
Refractive Index: 1.5350
Boiling Point: 114-116°/14mm
Beilstein Reference: 116243
MDL No.: MFCD00051668
Formula:
C₉H₁₂OS MW: 168.26 g/mol Boiling Pt: 114…116 °C/14 mm Density: 1,07 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051668 CAS Number: 20409-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...