3-Butenoic acid 96% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07557-14
L07557-22
CAAAAL07557-14EA
189.6
CAD
CAAAAL07557-14
CAAAAL07557-22
3-Butenoic acid 96% stabilized
3-Butenoic acid
Purity: 96%
CAS No.: 625-38-7
Molecular Formula: C4H6O2
Formula Weight: 86.09
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4240
Melting Point: -39°
Boiling Point: 162-163°
Beilstein Reference: 1699159
UN No.: 3265
MDL No.: MFCD00002782
Formula:
C₄H₆O₂ Boiling Pt: 162…163 °C Melting Pt: –39 °C Density: 1.012 Flash Pt: 65 °C(149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002782 CAS Number: 625-38-7 EINECS: 210-892-5 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...