N(α)-Cbz-L-tryptophan 98+%

Supplier: Thermo Scientific Chemicals

L07535-06 L07535-14
CAAAAL07535-06EA 40.45 CAD
CAAAAL07535-06 CAAAAL07535-14
N(α)-Cbz-L-tryptophan 98+%
N(α)-Cbz-L-tryptophan

Purity: 98+% CAS No.: 7432-21-5 Molecular Formula: C19H18N2O4 Formula Weight: 338.37 Storage Temperature: 2°C to 8°C Odor: Odorless Beilstein Reference: 96743 Optical Rotation: +3.1° (c=3 in acetic acid) MDL No.: MFCD00065700

Formula: C₁₉H₁₈N₂O₄
Melting Pt: 121…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065700
CAS Number: 7432-21-5
EINECS: 231-074-4

Order Now

Specification Test Results

Optical rotation +3.1° (c=3 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR