tert-Butylsulfonylacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

L07525-03 L07525-06
CAAAAL07525-03EA 115.34 CAD
CAAAAL07525-03 CAAAAL07525-06
tert-Butylsulfonylacetonitrile 98%
tert-Butylsulfonylacetonitrile

Purity: 98% CAS No.: 81536-18-7 Molecular Formula: C6H11NO2S Formula Weight: 161.22 Melting Point: 142-144° Beilstein Reference: 4370746 UN No.: 3439 MDL No.: MFCD00051622

Formula: C₆H₁₁NO₂S
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00051622
CAS Number: 81536-18-7
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR