4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%

Supplier: Thermo Scientific Chemicals

L07496-06 L07496-14
CAAAAL07496-06EA 62.73 CAD
CAAAAL07496-06 CAAAAL07496-14
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate

Purity: 98+% CAS No.: 388582-41-0 Molecular Formula: C5H7N3O.2H2O Formula Weight: 161.16 (125.13anhy) Melting Point: 300° Beilstein Reference: 118273 MDL No.: MFCD00150311

Formula: C₅H₇N₃O·2H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150311
CAS Number: 388582-41-0
EINECS: 212-182-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR