2-(Aminomethyl)benzimidazole dihydrochloride hydrate 98%

Supplier: Thermo Scientific Chemicals

L07440-06 L07440-14
CAAAAL07440-06EA 120.4 CAD
CAAAAL07440-06 CAAAAL07440-14
2-(Aminomethyl)benzimidazole dihydrochloride hydrate 98%
2-(Aminomethyl)benzimidazole dihydrochloride hydrate

Purity: 98% CAS No.: 5993-91-9 Molecular Formula: C8H9N3.2HCl.xH2O Formula Weight: 220.10(anhy) Melting Point: ca 264° dec. Beilstein Reference: 3698942 MDL No.: MFCD00150073

Formula: C₈H₉N₃·2HCl·xH₂O
MW: 220.10 g/mol (anhy)
Melting Pt: ∼ 264 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150073
CAS Number: 5993-91-9
EINECS: 227-820-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR