4,5-Dichloro-3(2H)-pyridazinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one
L07413-06
L07413-14
CAAAAL07413-06EA
55.77
CAD
CAAAAL07413-06
CAAAAL07413-14
4,5-Dichloro-3(2H)-pyridazinone 98%
4,5-Dichloro-3(2H)-pyridazinone
Purity: 98%
CAS No.: 932-22-9
Molecular Formula: C4H2Cl2N2O
Formula Weight: 164.98
Melting Point: 200-204°
Beilstein Reference: 120959
Formula:
C₄H₂Cl₂N₂O MW: 164.98 g/mol Melting Pt: 200…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051504 CAS Number: 932-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...