Het anhydride 95% may cont. up to 3% chlorendic acid
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorendic Anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride
L07349-0B
L07349-18
L07349-30
CAAAAL07349-0BEA
175.38
CAD
CAAAAL07349-0B
CAAAAL07349-18
CAAAAL07349-30
Het anhydride 95% may cont. up to 3% chlorendic acid
Het anhydride
Purity: 95%
CAS No.: 115-27-5
Molecular Formula: C9H2Cl6O3
Formula Weight: 370.83
Melting Point: 235-239°
Beilstein Reference: 92693
MDL No.: MFCD00080438
Formula:
C₉H₂Cl₆O₃ MW: 370.81 g/mol Melting Pt: 235…239 °C Density: 1.730 Storage Temperature: Ambient |
MDL Number:
MFCD00080438 CAS Number: 115-27-5 EINECS: 204-077-3 Merck Index: 14,2086 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...