D-(-)-α-(4-Hydroxyphenyl)glycine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine

L07190-06 L07190-14
CAAAAL07190-06EA 32.71 CAD
CAAAAL07190-06 CAAAAL07190-14
D-(-)-α-(4-Hydroxyphenyl)glycine 98+%
D(-)-α-(4-Hydroxyphenyl)glycine

Purity: 98+% CAS No.: 22818-40-2 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 240° dec. Beilstein Reference: 2210998

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004262
CAS Number: 22818-40-2
EINECS: 245-247-7

Order Now

Specification Test Results

Optical rotation -156° (c=1 in 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR