(Pyridin-2-ylsulfonyl)acetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07114-03
L07114-06
CAAAAL07114-03EA
135.09
CAD
CAAAAL07114-03
CAAAAL07114-06
(Pyridin-2-ylsulfonyl)acetonitrile 98%
(Pyridin-2-ylsulfonyl)acetonitrile
Purity: 98%
CAS No.: 170449-34-0
Molecular Formula: C7H6N2O2S
Formula Weight: 182.21
Melting Point: 77-79°
UN No.: 3439
MDL No.: MFCD00041443
Formula:
C₇H₆N₂O₂S Melting Pt: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041443 CAS Number: 170449-34-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...