Glycolaldehyde dimethyl acetal 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with sodium hydroxide 0.1%

L07099-03 L07099-06
CAAAAL07099-03EA 64.62 CAD
CAAAAL07099-03 CAAAAL07099-06
Glycolaldehyde dimethyl acetal 98% stabilized
Glycolaldehyde dimethyl acetal

Stab. with ca 0.1% sodium carbonate


Purity: 98% CAS No.: 30934-97-5 Molecular Formula: HOCH2CH(OCH3)2 Formula Weight: 106.12 Flash Point: 66°(150°F) Refractive Index: 1.4130 Melting Point: -76° Boiling Point: 68°/21mm Beilstein Reference: 1697583 MDL No.: MFCD00051799

Formula: HOCH₂CH(OCH₃)₂
Boiling Pt: 68 °C/21 mm
Melting Pt: –76 °C
Density: 1.050
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051799
CAS Number: 30934-97-5
EINECS: 250-398-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR