Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06941-03
L06941-MD
CAAAAL06941-03EA
224.36
CAD
CAAAAL06941-03
CAAAAL06941-MD
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 94%
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Purity: 94%
CAS No.: 175136-62-6
Molecular Formula: C24H9F18P
Formula Weight: 670.28
Melting Point: 98-102°
Boiling Point: 128-130°/0.5mm
MDL No.: MFCD00042428
Formula:
C₂₄H₉F₁₈P Boiling Pt: 128…130 °C/0.5 mm Melting Pt: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042428 CAS Number: 175136-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...