Carbonylcyanide-3-chlorophenylhydrazone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06932-MC
L06932-ME
CAAAAL06932-MCEA
42.98
CAD
CAAAAL06932-MC
CAAAAL06932-ME
Carbonylcyanide-3-chlorophenylhydrazone 98%
Carbonylcyanide-3-chlorophenylhydrazone
Purity: 98%
CAS No.: 555-60-2
Molecular Formula: C9H5ClN4
Formula Weight: 204.62
Storage Temperature: -30°C to -10°C
Odor: Odorless
Melting Point: ca 175° dec.
Beilstein Reference: 1842102
UN No.: 3439
MDL No.: MFCD00001848
Formula:
C₉H₅ClN₄ Melting Pt: ∼ 175 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00001848 CAS Number: 555-60-2 EINECS: 209-103-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...