Dimethylketene methyl trimethylsilyl acetal 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06931-06
L06931-14
CAAAAL06931-06EA
126.24
CAD
CAAAAL06931-06
CAAAAL06931-14
Dimethylketene methyl trimethylsilyl acetal 97%
Dimethylketene methyl trimethylsilyl acetal
Purity: 97%
CAS No.: 31469-15-5
Molecular Formula: C8H18O2Si
Formula Weight: 174.32
Flash Point: 30°(86°F)
Refractive Index: 1.4150
Boiling Point: 35-36°/15mm
Beilstein Reference: 1362893
UN No.: 1993
MDL No.: MFCD00010232
Formula:
C₈H₁₈O₂Si Boiling Pt: 35…36 °C/15 mm Density: 0.858 Flash Pt: 30 °C(86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010232 CAS Number: 31469-15-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...