3-Acetoxy-2-butanone 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
3-Oxobutan-2-yl acetate
L06927-06
L06927-14
CAAAAL06927-06EA
63.67
CAD
CAAAAL06927-06
CAAAAL06927-14
3-Acetoxy-2-butanone 98%
3-Acetoxy-2-butanone
Purity: 98%
CAS No.: 4906-24-5
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 66°(150°F)
Refractive Index: 1.4135
Boiling Point: 154-155°
Beilstein Reference: 1756882
MDL No.: MFCD00051782
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 154…155 °C Density: 1.025 Flash Pt: 66 °C(150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051782 CAS Number: 4906-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...