1-(2-Aminophenyl)pyrrole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(1H-Pyrrol-1-yl)aniline

L06887-03 L06887-06
CAAAAL06887-03EA 51.98 CAD
CAAAAL06887-03 CAAAAL06887-06
1-(2-Aminophenyl)pyrrole 98+%
1-(2-Aminophenyl)pyrrole

Purity: 98+% CAS No.: 6025-60-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 93-94° Beilstein Reference: 1307631

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005344
CAS Number: 6025-60-1
EINECS: 227-884-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR