2-(Methylthio)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06870-03
L06870-06
CAAAAL06870-03EA
104.28
CAD
CAAAAL06870-03
CAAAAL06870-06
2-(Methylthio)benzonitrile 98%
2-(Methylthio)benzonitrile
Purity: 98%
CAS No.: 6609-54-7
Molecular Formula: C8H7NS
Formula Weight: 149.22
Melting Point: 33-38°
Boiling Point: 116-117°/5mm
UN No.: 3439
MDL No.: MFCD00015557
Formula:
C₈H₇NS Boiling Pt: 116…117 °C/5 mm Melting Pt: 33…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015557 CAS Number: 6609-54-7 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...