Nɑ-Acetyl-L-glutamine 99%
Supplier: Thermo Scientific Chemicals
L06780-18
L06780-30
CAAAAL06780-18EA
74.73
CAD
CAAAAL06780-18
CAAAAL06780-30
Nɑ-Acetyl-L-glutamine 99%
Nalpha-Acetyl-L-glutamine
Purity: 99%
CAS No.: 2490-97-3
Molecular Formula: C7H12N2O4
Formula Weight: 188.18
Storage Temperature: 2°C to 8°C
Melting Point: ca 206° dec.
Beilstein Reference: 1727471
Optical Rotation: -12° (c=3 in water)
MDL No.: MFCD00038159
Formula:
C₇H₁₂N₂O₄ Melting Pt: ca 206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038159 CAS Number: 2490-97-3 EINECS: 219-647-7 Merck Index: 14,25 |
Specification Test Results
Optical rotation | -12° (c=3 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...