6-Azathymine 98%
Supplier: Thermo Scientific Chemicals
L06762-06
L06762-14
CAAAAL06762-06EA
184.86
CAD
CAAAAL06762-06
CAAAAL06762-14
6-Azathymine 98%
6-Azathymine
Purity: 98%
CAS No.: 932-53-6
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point: 211-215°
Beilstein Reference: 126863
MDL No.: MFCD00006457
Formula:
C₄H₅N₃O₂ Melting Pt: 211…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006457 CAS Number: 932-53-6 EINECS: 213-253-9 Merck Index: 14,903 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...