3,5-Pyrazoledicarboxylic acid monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Pyrazole-3,5-dicarboxylic acid hydrate, Pyrazole-3,5-dicarboxylic acid monohydrate, 1H-Pyrazole-3,5-dicarboxylic acid monohydrate

L06731-06 L06731-14 L06731-22
CAAAAL06731-06EA 118.18 CAD
CAAAAL06731-06 CAAAAL06731-14 CAAAAL06731-22
3,5-Pyrazoledicarboxylic acid monohydrate 98%
3,5-Pyrazoledicarboxylic acid monohydrate

Purity: 98% CAS No.: 303180-11-2 Molecular Formula: C5H4N2O4.H2O Formula Weight: 174.11 (156.09anhy) Melting Point: ca 294° dec. Beilstein Reference: 147821 MDL No.: MFCD00149323

Formula: C₅H₄N₂O₄·H₂O
MW: 174.11 g/mol
Melting Pt: ca 294 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149323
CAS Number: 303180-11-2
EINECS: 221-474-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR