Dimethyl-3-nitrophthalate 98%
Supplier: Thermo Scientific Chemicals
L06715-06
L06715-14
CAAAAL06715-06EA
108.39
CAD
CAAAAL06715-06
CAAAAL06715-14
Dimethyl-3-nitrophthalate 98%
Dimethyl 3-nitrophthalate
Purity: 98%
CAS No.: 13365-26-9
Molecular Formula: C10H9NO6
Formula Weight: 239.19
Melting Point: 67-70°
Beilstein Reference: 1885570
Formula:
C₁₀H₉NO₆ Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017184 CAS Number: 13365-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...