1-Naphthylacetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06696-06
L06696-14
L06696-22
CAAAAL06696-06EA
36.97
CAD
CAAAAL06696-06
CAAAAL06696-14
CAAAAL06696-22
1-Naphthylacetonitrile 97%
1-Naphthylacetonitrile
Purity: 97%
CAS No.: 132-75-2
Molecular Formula: C12H9N
Formula Weight: 167.21
Flash Point:
Formula:
C₁₂H₉N Boiling Pt: 180…182 °C/9 mm Melting Pt: 33…35 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004041 CAS Number: 132-75-2 EINECS: 205-078-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...