Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate

L06654-03 L06654-06 L06654-14
CAAAAL06654-03EA 39.66 CAD
CAAAAL06654-03 CAAAAL06654-06 CAAAAL06654-14
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate

Purity: 95% CAS No.: 15400-53-0 Molecular Formula: C7H9N3O3 Formula Weight: 183.17 Odor: Odorless Melting Point: ca 300° dec. Beilstein Reference: 165835 MDL No.: MFCD00039708

Formula: C₇H₉N₃O₃
MW: 183.17 g/mol
Melting Pt: ∼ 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00039708
CAS Number: 15400-53-0
EINECS: 239-418-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR