5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: EDANS sodium salt

L06555-03 L06555-MD
CAAAAL06555-03EA 241.74 CAD
CAAAAL06555-03 CAAAAL06555-MD
5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt 97%
5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt

Purity: 97% CAS No.: 100900-07-0 Molecular Formula: C12H13N2NaO3S Formula Weight: 288.30 Melting Point: 300° Beilstein Reference: 4283850 MDL No.: MFCD00051474

Formula: C₁₂H₁₃N₂NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00051474
CAS Number: 100900-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR