DL-Valine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Val-OMe.HCl, H-DL-Val-OMe·HCl

L06526-06 L06526-14
CAAAAL06526-06EA 74.58 CAD
CAAAAL06526-06 CAAAAL06526-14
DL-Valine methyl ester hydrochloride 99%
DL-Valine methyl ester hydrochloride

Purity: 99% CAS No.: 5619-05-6 Molecular Formula: C6H13NO2.HCl Formula Weight: 167.64 Melting Point: 112-114° Beilstein Reference: 3912090 MDL No.: MFCD00066118

Formula: C₆H₁₃NO₂
MW: 167.63 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00066118
CAS Number: 5619-05-6
EINECS: 227-048-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR