2,2'-Dichlorobenzil ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,2-Bis(2-chlorophenyl)ethane-1,2-dione
L06507-06
L06507-14
CAAAAL06507-06EA
137.14
CAD
CAAAAL06507-06
CAAAAL06507-14
2,2'-Dichlorobenzil ≥98%
2,2'-Dichlorobenzil
Purity: 98+%
CAS No.: 21854-95-5
Molecular Formula: C14H8Cl2O2
Formula Weight: 279.12
Melting Point: 132-136°
Beilstein Reference: 2120689
MDL No.: MFCD00018263
Formula:
C₁₄H₈Cl₂O₂ MW: 279.12 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018263 CAS Number: 21854-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...