1-(2-Hydroxyethyl)hexamethylenimine 95%

Supplier: Thermo Scientific Chemicals
Danger

L06497-06 L06497-14
CAAAAL06497-06EA 60.2 CAD
CAAAAL06497-06 CAAAAL06497-14
1-(2-Hydroxyethyl)hexamethylenimine 95%
1-(2-Hydroxyethyl)hexamethylenimine

Purity: 95% CAS No.: 20603-00-3 Molecular Formula: C8H17NO Formula Weight: 143.22 Refractive Index: 1.4840 Boiling Point: 114-115°/23mm Beilstein Reference: 104110 UN No.: 2735 MDL No.: MFCD00020988

Formula: C₈H₁₇NO
Boiling Pt: 114…115 °C/23 mm
Density: 0.971
Storage Temperature: Ambient
MDL Number: MFCD00020988
CAS Number: 20603-00-3
EINECS: 243-915-2
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR