5-Chloro-m-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06485-06
L06485-14
CAAAAL06485-06EA
225.94
CAD
CAAAAL06485-06
CAAAAL06485-14
5-Chloro-m-phenylenediamine 97%
5-Chloro-m-phenylenediamine
Purity: 97%
CAS No.: 33786-89-9
Molecular Formula: C6H7ClN2
Formula Weight: 142.59
Melting Point: 99-103°
Beilstein Reference: 3029854
UN No.: 1673
MDL No.: MFCD00014775
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014775 CAS Number: 33786-89-9 UN: 1673 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...