1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L06476-09 L06476-18
CAAAAL06476-04EA 97.79 CAD
CAAAAL06476-04 CAAAAL06476-09 CAAAAL06476-18
1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine ≥98%
1-Phenyl-4,5-dihydro-1H-pyrazol-3-ylamine

Purity: 98+% CAS No.: 3314-35-0 Molecular Formula: C9H11N3 Formula Weight: 161.21 Melting Point: 167-170° Beilstein Reference: 139162 MDL No.: MFCD00051730

Formula: C₉H₁₁N₃
MW: 161.21 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00051730
CAS Number: 3314-35-0
EINECS: 222-005-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR