Ethyl-4-hydroxy-3-nitrobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06451-06
L06451-14
CAAAAL06451-06EA
61.62
CAD
CAAAAL06451-06
CAAAAL06451-14
Ethyl-4-hydroxy-3-nitrobenzoate 98%
Ethyl-4-hydroxy-3-nitrobenzoate
Purity: 98%
CAS No.: 19013-10-6
Molecular Formula: C9H9NO5
Formula Weight: 211.18
Melting Point: 69-71°
Beilstein Reference: 2121319
MDL No.: MFCD00016999
Formula:
C₉H₉NO₅ Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016999 CAS Number: 19013-10-6 EINECS: 242-751-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...