2-Chlorophenyl cyclopentyl ketone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Chlorobenzoylcyclopentane

L06448-09 L06448-18
CAAAAL06448-09EA 153.1 CAD
CAAAAL06448-09 CAAAAL06448-18
2-Chlorophenyl cyclopentyl ketone 97%
2-Chlorophenyl cyclopentyl ketone

Purity: 97% CAS No.: 6740-85-8 Molecular Formula: C12H13ClO Formula Weight: 208.69 Flash Point: 100°(212°F) Refractive Index: 1.5470 Odor: Aromatic Boiling Point: 130°/5mm Beilstein Reference: 1873022 MDL No.: MFCD00038367

Formula: C₁₂H₁₃ClO
Boiling Pt: 130 °C/5 mm
Density: 1.16
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038367
CAS Number: 6740-85-8
EINECS: 229-802-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR